Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286055.1 | NP_001272984.1 | 5 | 730 | ||
NM_001286056.1 | NP_001272985.1 | 6 | 658 | |||
NM_001839.4 | NP_001830.1 | 6 | 868 | |||
XM_017000245.1 | XP_016855734.1 | 8 | 3280 | |||
GenBank mRNA | AK294141.1 | - | 6 | 619 | ||
AK298145.1 | - | 5 | 650 | |||
AK312370.1 | - | 6 | 755 | |||
AK316398.1 | - | 6 | 634 | |||
BC025372.2 | - | 6 | 752 | 3505668 | ||
CR541848.1 | - | 6 | 539 | |||
DR157009.1 | - | 6 | 791 | |||
S80562.1 | - | 6 | 622 |