Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSF2RA SIRNA Assays ›
Gene Symbol:
CSF2RA
Gene Name:
colony stimulating factor 2 receptor alpha subunit
Gene Aliases:
CD116, CDw116, CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF-R-alpha, GMCSFR, GMR, SMDP4
Chromosome Location:
Chr.X: 1268800 - 1325097 on Build GRCh38
UniGene ID:
Hs.520937
Species:
Human
siRNA ID
HSS102362
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSF2RA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001161529.1
NP_001155001.1
8
878
NM_001161530.1
NP_001155002.1
5
582
NM_001161531.1
NP_001155003.1
7
750
NM_001161532.1
NP_001155004.1
5
519
NM_006140.4
NP_006131.2
7
750
NM_172245.2
NP_758448.1
7
726
NM_172246.2
NP_758449.1
7
750
NM_172247.2
NP_758450.1
5
582
NM_172249.2
NP_758452.1
7
750
NR_027760.1
7
750
XM_011546165.2
XP_011544467.1
6
628
XM_011546166.1
XP_011544468.1
6
628
XM_011546167.2
XP_011544469.1
6
628
XM_011546168.2
XP_011544470.1
6
628
XM_011546169.1
XP_011544471.1
6
628
XM_011546170.2
XP_011544472.1
6
628
XM_011546173.1
XP_011544475.1
6
628
XM_011546174.2
XP_011544476.1
6
628
XM_011546175.2
XP_011544477.1
6
628
XM_017029287.1
XP_016884776.1
7
740
XM_017029288.1
XP_016884777.1
7
717
GenBank mRNA
AK293086.1
-
7
708
AK301395.1
-
5
501
AK309630.1
-
7
711
AL833597.1
-
7
687
BC002635.1
-
7
726
3606186
DQ841258.1
-
5
556
L29348.1
-
7
705
L29349.1
-
7
705
M64445.1
-
7
718
M73832.1
-
5
574
U93096.1
-
7
705
X17648.1
-
7
705
X54935.1
-
5
556
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
peptidyl-tyrosine phosphorylation
granulocyte colony-stimulating factor signaling pathway
positive regulation of GTPase activity
cellular protein metabolic process
cellular response to granulocyte macrophage colony-stimulating factor stimulus
protein tyrosine kinase activity
receptor activity
granulocyte colony-stimulating factor receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
granulocyte colony-stimulating factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0