Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DGKA SIRNA Assays ›
Gene Symbol:
DGKA
Gene Name:
diacylglycerol kinase alpha
Gene Aliases:
DAGK, DAGK1, DGK-alpha
Chromosome Location:
Chr.12: 55931076 - 55954023 on Build GRCh38
UniGene ID:
Hs.524488
Species:
Human
siRNA ID
HSS102626
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DGKA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001345.4
NP_001336.2
15
1481
NM_201444.2
NP_958852.1
15
1526
NM_201445.1
NP_958853.1
16
1494
NM_201554.1
NP_963848.1
15
1394
XM_005268688.2
XP_005268745.1
15
1423
XM_005268689.2
XP_005268746.1
15
1358
XM_005268690.2
XP_005268747.1
13
1178
XM_011537991.2
XP_011536293.1
16
1591
XM_011537993.2
XP_011536295.1
15
1342
XM_011537995.2
XP_011536297.1
15
1353
XM_017018900.1
XP_016874389.1
14
1777
XM_017018901.1
XP_016874390.1
14
1921
XM_017018902.1
XP_016874391.1
14
1743
XM_017018903.1
XP_016874392.1
15
1945
XM_017018904.1
XP_016874393.1
15
1813
XM_017018905.1
XP_016874394.1
16
2222
XM_017018906.1
XP_016874395.1
14
1854
XM_017018907.1
XP_016874396.1
14
1677
XM_017018908.1
XP_016874397.1
15
1561
XM_017018909.1
XP_016874398.1
14
1312
GenBank mRNA
AF064767.1
-
1
83
AF064769.1
-
15
1403
AF064770.1
-
15
1471
AF064771.1
-
15
1439
AK122973.1
-
13
2832
AK127093.1
-
7
2169
AK226134.1
-
9
3314
AK307685.1
-
14
1238
AY930112.1
-
14
1332
BC023523.2
-
16
1417
4304187
BC031870.2
-
15
1421
4819985
X62535.1
-
14
1320
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidic acid biosynthetic process
protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
platelet activation
intracellular signal transduction
diacylglycerol kinase activity
calcium ion binding
ATP binding
phospholipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0