Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171184.1 | NP_001164655.1 | 20 | 2609 | ||
NM_001939.2 | NP_001930.2 | 22 | 2892 | |||
XM_017029333.1 | XP_016884822.1 | 21 | 2975 | |||
XM_017029334.1 | XP_016884823.1 | 21 | 2971 | |||
GenBank mRNA | AB209305.1 | - | 13 | 1489 | ||
AK289825.1 | - | 22 | 2780 | |||
AK295843.1 | - | 20 | 2478 | |||
BC111695.1 | - | 22 | 2729 | 40036070 | ||
U43519.1 | - | 22 | 2841 |