Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004446.2 | NP_004437.2 | 9 | 1367 | ||
GenBank mRNA | AK296755.1 | - | 9 | 1314 | ||
AK307534.1 | - | 7 | 941 | |||
AY493416.1 | - | 7 | 914 | |||
BC034797.1 | - | 9 | 1163 | |||
BC058921.1 | - | 9 | 1204 | |||
BC067841.1 | - | 8 | 1144 | |||
BC105983.1 | - | 9 | 1165 | |||
BC126275.1 | - | 9 | 1235 | 8991991 | ||
BC136465.1 | - | 9 | 1135 | 9020454 | ||
CR933648.1 | - | 9 | 1206 | |||
JQ080075.1 | - | 9 | 1097 | |||
X07466.1 | - | 5 | 597 | |||
X54326.1 | - | 7 | 939 |