Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ESRRG SIRNA Assays ›
Gene Symbol:
ESRRG
Gene Name:
estrogen related receptor gamma
Gene Aliases:
ERR3, ERRgamma, NR3B3
Chromosome Location:
Chr.1: 216503246 - 217137757 on Build GRCh38
UniGene ID:
Hs.444225
Species:
Human
siRNA ID
HSS103388
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ESRRG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134285.2
NP_001127757.1
8
1362
NM_001243505.1
NP_001230434.1
6
802
NM_001243506.1
NP_001230435.1
6
872
NM_001243507.1
NP_001230436.1
7
1649
NM_001243509.1
NP_001230438.1
8
1740
NM_001243510.2
NP_001230439.1
8
1356
NM_001243511.2
NP_001230440.1
7
1238
NM_001243512.1
NP_001230441.1
8
1305
NM_001243513.1
NP_001230442.1
7
1325
NM_001243514.1
NP_001230443.1
6
1482
NM_001243515.1
NP_001230444.1
8
1581
NM_001243518.1
NP_001230447.1
7
1435
NM_001243519.1
NP_001230448.1
8
1585
NM_001438.3
NP_001429.2
6
1286
NM_206594.2
NP_996317.1
8
1406
NM_206595.2
NP_996318.1
7
1288
XM_011509265.2
XP_011507567.1
8
5370
XM_011509266.2
XP_011507568.1
8
1743
XM_011509267.1
XP_011507569.1
9
1687
XM_011509268.2
XP_011507570.1
8
1572
XM_011509269.2
XP_011507571.1
6
1171
XM_011509270.1
XP_011507572.1
6
1459
XM_011509271.2
XP_011507573.1
7
1452
XM_011509274.1
XP_011507576.1
8
1480
XM_011509275.1
XP_011507577.1
8
1436
XM_011509276.1
XP_011507578.1
8
1595
XM_011509277.1
XP_011507579.1
7
1287
XM_011509278.1
XP_011507580.1
7
1318
XM_011509279.1
XP_011507581.1
7
1463
XM_011509280.2
XP_011507582.1
7
1440
XM_017000621.1
XP_016856110.1
9
5528
XM_017000622.1
XP_016856111.1
8
1760
XM_017000623.1
XP_016856112.1
8
1720
XM_017000624.1
XP_016856113.1
8
4086
XM_017000625.1
XP_016856114.1
8
1623
XM_017000626.1
XP_016856115.1
8
1480
XM_017000627.1
XP_016856116.1
8
1543
XM_017000628.1
XP_016856117.1
10
1772
XM_017000629.1
XP_016856118.1
7
1316
XM_017000630.1
XP_016856119.1
10
1818
XM_017000631.1
XP_016856120.1
9
1565
XM_017000632.1
XP_016856121.1
10
1744
XM_017000633.1
XP_016856122.1
8
1405
XM_017000634.1
XP_016856123.1
8
1447
XM_017000635.1
XP_016856124.1
10
4522
XM_017000636.1
XP_016856125.1
9
1591
XM_017000637.1
XP_016856126.1
9
1692
XM_017000638.1
XP_016856127.1
9
1544
XM_017000639.1
XP_016856128.1
8
1384
XM_017000640.1
XP_016856129.1
7
3647
XM_017000641.1
XP_016856130.1
7
1442
XM_017000642.1
XP_016856131.1
8
1540
XM_017000643.1
XP_016856132.1
9
1537
XM_017000644.1
XP_016856133.1
8
1415
XM_017000645.1
XP_016856134.1
7
1297
XM_017000646.1
XP_016856135.1
8
3780
XM_017000647.1
XP_016856136.1
7
1529
XM_017000648.1
XP_016856137.1
7
1418
XM_017000649.1
XP_016856138.1
9
1866
GenBank mRNA
AB020639.1
-
6
1267
AB307715.1
-
5
1044
AB362218.1
-
8
1659
AF058291.1
-
7
1262
AF094518.1
-
6
1238
AK131193.1
-
7
1610
AK290945.1
-
8
1562
AK291028.1
-
7
1263
AK291647.1
-
6
1259
AK298851.1
-
6
839
AY388456.1
-
6
1241
AY388457.1
-
7
1274
AY388458.1
-
8
1300
AY388459.1
-
7
1317
AY388460.1
-
6
1157
AY388461.1
-
6
1531
BC064700.1
-
7
1416
30344716
BX647076.1
-
8
1362
CR749497.1
-
8
1355
HQ692853.1
-
6
1113
HQ692854.1
-
5
1044
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
steroid hormone mediated signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
steroid hormone receptor activity
retinoic acid receptor activity
steroid binding
protein binding
zinc ion binding
AF-2 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0