Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004486.4 | NP_004477.3 | 15 | 1196 | ||
XM_005251930.1 | XP_005251987.1 | 16 | 1295 | |||
XM_005251931.1 | XP_005251988.1 | 15 | 1214 | |||
XM_005251932.1 | XP_005251989.1 | 14 | 1126 | |||
GenBank mRNA | AF248953.1 | - | 15 | 1147 | ||
BC014188.2 | - | 3 | 213 | |||
BC069268.1 | - | 15 | 1196 | 6340972 | ||
L06147.1 | - | 1 | 55 |