Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270927.1 | NP_001257856.1 | 3 | 384 | ||
NM_001270928.1 | NP_001257857.1 | 3 | 401 | |||
NM_001270929.1 | NP_001257858.1 | 3 | 396 | |||
NM_001270930.1 | NP_001257859.1 | 3 | 522 | |||
NM_001548.4 | NP_001539.3 | 2 | 287 | |||
GenBank mRNA | AK092813.1 | - | 2 | 1315 | ||
AK308688.1 | - | 3 | 288 | |||
AK314588.1 | - | 2 | 210 | |||
AL548182.3 | - | 2 | 190 | |||
AL711085.1 | - | 3 | 297 | |||
BC007091.1 | - | 2 | 212 | 4250452 | ||
BI670242.1 | - | 2 | 295 | |||
BT006667.1 | - | 1 | 120 | |||
CD106320.1 | - | 2 | 163 | |||
DA822484.1 | - | 2 | 269 | |||
DB003897.1 | - | 3 | 321 | |||
DB180094.1 | - | 3 | 331 | |||
X03557.1 | - | 2 | 184 |