Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270927.1 | NP_001257856.1 | 3 | 513 | ||
NM_001270928.1 | NP_001257857.1 | 3 | 530 | |||
NM_001270929.1 | NP_001257858.1 | 3 | 525 | |||
NM_001270930.1 | NP_001257859.1 | 3 | 651 | |||
NM_001548.4 | NP_001539.3 | 2 | 416 | |||
GenBank mRNA | AK092813.1 | - | 2 | 1444 | ||
AK308688.1 | - | 3 | 417 | |||
AK314588.1 | - | 2 | 339 | |||
AL548182.3 | - | 2 | 319 | |||
AL711085.1 | - | 3 | 426 | |||
BC007091.1 | - | 2 | 341 | 4250452 | ||
BI670242.1 | - | 2 | 424 | |||
BT006667.1 | - | 1 | 249 | |||
CD106320.1 | - | 2 | 292 | |||
DA822484.1 | - | 2 | 398 | |||
DB003897.1 | - | 3 | 450 | |||
DB180094.1 | - | 3 | 460 | |||
X03557.1 | - | 2 | 313 |