Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL7R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002185.3 | NP_002176.2 | 5 | 763 | ||
XM_005248299.3 | XP_005248356.1 | 5 | 748 | |||
GenBank mRNA | AK301220.1 | - | 5 | 747 | ||
AK315251.1 | - | 5 | 662 | |||
BC020717.1 | - | 5 | 749 | |||
BC067537.1 | - | 5 | 740 | 6971840 | ||
BC067538.1 | - | 5 | 740 | 6971842 | ||
BC067539.1 | - | 5 | 740 | 6971844 | ||
BC067540.1 | - | 5 | 740 | 6971846 | ||
BC069999.1 | - | 5 | 740 | 6971843 | ||
M29696.1 | - | 5 | 682 |