Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001075098.3 | NP_001068566.1 | 11 | 1516 | ||
NM_005943.5 | NP_005934.2 | 10 | 1520 | |||
NR_033233.1 | 9 | 1326 | ||||
XM_011514632.2 | XP_011512934.1 | 11 | 1412 | |||
XM_011514633.2 | XP_011512935.1 | 10 | 1364 | |||
XM_011514634.2 | XP_011512936.1 | 10 | 1309 | |||
XM_017010888.1 | XP_016866377.1 | 10 | 1334 | |||
GenBank mRNA | AF034374.1 | - | 11 | 1399 | ||
AF214015.1 | - | 5 | 536 | |||
AF214022.1 | - | 2 | 240 | |||
AF214023.1 | - | 4 | 473 | |||
AJ224328.1 | - | 10 | 1517 | |||
AK056740.1 | - | 9 | 1272 | |||
AK091306.1 | - | 10 | 1248 | |||
AY423726.1 | - | 1 | 212 | |||
CX787499.1 | - | 7 | 782 |