Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257102.1 | NP_001244031.1 | 3, 4 | 492 | ||
NM_002491.2 | NP_002482.1 | 2, 3 | 469 | |||
XM_011511230.2 | XP_011509532.1 | 3, 4 | 506 | |||
XM_017004186.1 | XP_016859675.1 | 2, 3 | 714 | |||
GenBank mRNA | AF035839.1 | - | 2, 3 | 294 | ||
AF047183.1 | - | 2, 3 | 398 | |||
BC018183.1 | - | 2, 3 | 417 | 3881592 | ||
BM848639.1 | - | 2, 3 | 469 | |||
BU188560.1 | - | 2, 3 | 196 | |||
CR456924.1 | - | 1, 2 | 146 |