Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021074.4 | NP_066552.2 | 8 | 808 | ||
XM_017025782.1 | XP_016881271.1 | 8 | 998 | |||
GenBank mRNA | AF147367.1 | - | 4 | 283 | ||
AK291865.1 | - | 8 | 704 | |||
AK312129.1 | - | 8 | 726 | |||
AW051350.1 | - | 1 | 126 | |||
BC001632.1 | - | 8 | 718 | 3536345 | ||
BC017487.1 | - | 8 | 716 | 3537815 | ||
BC071689.1 | - | 8 | 698 | 4372795 | ||
BG742948.1 | - | 8 | 808 | |||
CR456928.1 | - | 8 | 694 | |||
M22538.1 | - | 8 | 712 |