Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003941.2 | NP_001003941.1 | 9 | 1324 | ||
NM_001165036.1 | NP_001158508.1 | 9 | 1312 | |||
NM_002541.3 | NP_002532.2 | 9 | 1324 | |||
XM_005249759.4 | XP_005249816.1 | 10 | 1301 | |||
XM_005249761.3 | XP_005249818.1 | 10 | 1307 | |||
XM_011515408.2 | XP_011513710.1 | 11 | 1388 | |||
GenBank mRNA | AK293871.1 | - | 9 | 1080 | ||
AK294951.1 | - | 6 | 698 | |||
AK296400.1 | - | 6 | 825 | |||
AK302805.1 | - | 7 | 936 | |||
AK304439.1 | - | 9 | 1243 | |||
AK304690.1 | - | 7 | 907 | |||
AK314179.1 | - | 9 | 1246 | |||
BC004964.1 | - | 8 | 1207 | 3528538 | ||
BC009580.1 | - | 9 | 1258 | |||
BC014617.2 | - | 8 | 1207 | 3010095 | ||
BC128549.1 | - | 8 | 871 | |||
BC128550.1 | - | 8 | 883 | |||
BM998792.1 | - | 1 | 503 | |||
D10523.1 | - | 9 | 1222 |