Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAWR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002583.2 | NP_002574.2 | 6 | 1160 | ||
XM_006719435.2 | XP_006719498.1 | 6 | 1136 | |||
XM_017019377.1 | XP_016874866.1 | 6 | 1040 | |||
GenBank mRNA | BC007018.1 | - | 5 | 1006 | 3828412 | |
CR536549.1 | - | 5 | 874 | |||
U63809.1 | - | 6 | 1118 |