Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PFKM SIRNA Assays ›
Gene Symbol:
PFKM
Gene Name:
phosphofructokinase, muscle
Gene Aliases:
ATP-PFK, GSD7, PFK-1, PFK1, PFKA, PFKX, PPP1R122
Chromosome Location:
Chr.12: 48105278 - 48146404 on Build GRCh38
UniGene ID:
Hs.75160
Species:
Human
siRNA ID
HSS107873
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PFKM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000289.5
NP_000280.1
14
1454
NM_001166686.1
NP_001160158.1
16
1756
NM_001166687.1
NP_001160159.1
14
1308
NM_001166688.1
NP_001160160.1
13
1397
XM_005268974.1
XP_005269031.1
17
1653
XM_005268975.1
XP_005269032.1
17
1792
XM_005268976.3
XP_005269033.1
17
1835
XM_005268977.1
XP_005269034.1
16
1639
XM_005268978.2
XP_005269035.1
16
1552
XM_005268979.1
XP_005269036.1
16
1639
XM_011538487.1
XP_011536789.1
15
1494
XM_011538488.2
XP_011536790.1
14
1419
XM_017019467.1
XP_016874956.1
17
1722
XM_017019468.1
XP_016874957.1
16
1574
XM_017019469.1
XP_016874958.1
15
1546
GenBank mRNA
AK126020.1
-
18
2109
AK126229.1
-
17
1706
AK303681.1
-
9
801
AK308185.1
-
9
777
AK312284.1
-
14
1336
BC000534.2
-
13
1284
2964710
BC007798.2
-
12
1171
BC012799.2
-
13
1284
2964710
BC013298.2
-
13
1284
2964710
BC021203.2
-
14
1306
4100117
CR541698.1
-
4
324
J05533.1
-
5
365
KU178251.1
-
13
1269
KU178252.1
-
12
1176
KU178253.1
-
13
1269
KU178254.1
-
2
114
M26066.1
-
14
1345
U24183.1
-
13
1597
Y00698.1
-
13
1288
Back To Top
More Information
Panther Classification:
Molecular Function -
carbohydrate kinase
kinase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
glycogen catabolic process
fructose 6-phosphate metabolic process
glycolytic process
positive regulation of insulin secretion
glucose homeostasis
muscle cell cellular homeostasis
carbohydrate phosphorylation
protein oligomerization
canonical glycolysis
glycolysis from storage polysaccharide through glucose-1-phosphate
6-phosphofructokinase activity
protein binding
ATP binding
protein C-terminus binding
kinase binding
identical protein binding
protein homodimerization activity
metal ion binding
fructose binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0