Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021129.3 | NP_066952.1 | 4 | 353 | ||
GenBank mRNA | AB026723.1 | - | 4 | 276 | ||
AF092439.1 | - | 4 | 323 | |||
AF108211.1 | - | 4 | 234 | |||
AF119665.1 | - | 4 | 323 | |||
AF154065.1 | - | 4 | 291 | |||
AF217186.1 | - | 4 | 246 | |||
AK058193.1 | - | 4 | 346 | |||
AY295767.1 | - | 1 | 442 | |||
BC001022.2 | - | 4 | 328 | 3345496 | ||
BC061581.1 | - | 4 | 336 | 6526628 | ||
BC105034.1 | - | 4 | 319 | 8144037 | ||
BC105036.1 | - | 4 | 319 | 8144039 | ||
BC107882.1 | - | 4 | 287 | 6495107 | ||
BU520836.1 | - | 4 | 334 | |||
EU794626.1 | - | 4 | 353 |