Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206994.1 | NP_001193923.1 | 10 | 1155 | ||
NM_001206995.1 | NP_001193924.1 | 10 | 1259 | |||
NM_001206996.1 | NP_001193925.1 | 10 | 1309 | |||
NM_020416.3 | NP_065149.2 | 9 | 1447 | |||
NM_181876.2 | NP_870991.1 | 9 | 1101 | |||
XM_005247979.4 | XP_005248036.1 | 5 | 1292 | |||
XM_011513495.1 | XP_011511797.1 | 9 | 1196 | |||
XM_011513496.1 | XP_011511798.1 | 9 | 1211 | |||
GenBank mRNA | AF086924.1 | - | 9 | 1131 | ||
AF145027.1 | - | 9 | 1294 | |||
AK093115.1 | - | 9 | 1101 | |||
AK122849.1 | - | 5 | 3108 | |||
AK296474.1 | - | 10 | 1155 | |||
AK316003.1 | - | 11 | 1269 | |||
BC032954.2 | - | 10 | 1309 | 4823280 |