Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006265.2 | NP_006256.1 | 6 | 877 | ||
GenBank mRNA | AK098521.1 | - | 2 | 121 | ||
AK289505.1 | - | 6 | 654 | |||
BC050381.1 | - | 6 | 717 | 6044010 | ||
D38551.1 | - | 6 | 773 | |||
X98294.1 | - | 5 | 589 |