Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291446.1 | NP_001278375.1 | 2 | 521 | ||
NM_001291447.1 | NP_001278376.1 | 2 | 501 | |||
NM_005407.2 | NP_005398.2 | 2 | 521 | |||
NR_111966.1 | 2 | 521 | ||||
NR_111967.1 | 2 | 501 | ||||
XM_011537064.1 | XP_011535366.1 | 2 | 447 | |||
XM_011537065.1 | XP_011535367.1 | 2 | 593 | |||
GenBank mRNA | AB002358.2 | - | 2 | 508 | ||
AK001276.1 | - | 2 | 239 | |||
AK289358.1 | - | 2 | 337 | |||
AK293574.1 | - | 2 | 207 | |||
AK294051.1 | - | 2 | 302 | |||
AK296414.1 | - | 2 | 320 | |||
AK299931.1 | - | 2 | 395 | |||
BC024245.2 | - | 2 | 314 | |||
BC090958.1 | - | 2 | 321 | |||
BC136528.1 | - | 2 | 256 | 9020518 | ||
BC136529.1 | - | 2 | 256 | 9020519 | ||
BI823697.1 | - | 2 | 321 | |||
DC306696.1 | - | 2 | 521 | |||
DC307767.1 | - | 2 | 333 | |||
DC309911.1 | - | 2 | 501 | |||
DT216192.1 | - | 2 | 197 | |||
X98834.1 | - | 2 | 408 |