Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284351.2 | NP_001271280.1 | 4, 5 | 862 | ||
NM_004174.3 | NP_004165.2 | 4, 5 | 862 | |||
GenBank mRNA | BC101669.1 | - | 4, 5 | 758 | 8069175 | |
BC101671.1 | - | 4, 5 | 758 | 8069177 | ||
BC143328.1 | - | 4, 5 | 758 | 9051833 | ||
U28043.1 | - | 4, 5 | 760 |