Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TROVE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042369.2 | NP_001035828.1 | 8 | 1755 | ||
NM_001042370.2 | NP_001035829.2 | 8 | 1755 | |||
NM_001173524.1 | NP_001166995.1 | 8 | 1761 | |||
NM_001173525.1 | NP_001166996.1 | 8 | 1755 | |||
NM_004600.5 | NP_004591.2 | 8 | 1755 | |||
NR_033393.1 | 7 | 1160 | ||||
XM_006711495.3 | XP_006711558.1 | 9 | 1827 | |||
XM_006711496.3 | XP_006711559.1 | 10 | 2003 | |||
XM_006711497.3 | XP_006711560.1 | 9 | 1937 | |||
XM_011509922.2 | XP_011508224.1 | 8 | 1336 | |||
XM_017002180.1 | XP_016857669.1 | 9 | 1636 | |||
XM_017002181.1 | XP_016857670.1 | 9 | 1865 | |||
XM_017002182.1 | XP_016857671.1 | 8 | 1570 | |||
XM_017002183.1 | XP_016857672.1 | 6 | 907 | |||
GenBank mRNA | AK314594.1 | - | 8 | 1548 | ||
AY205314.1 | - | 8 | 1569 | |||
AY205315.1 | - | 8 | 1569 | |||
BC036658.2 | - | 8 | 1597 | 4821776 | ||
CR936683.1 | - | 7 | 1129 | |||
J04137.1 | - | 7 | 1424 | |||
M25077.1 | - | 8 | 1569 |