Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003162.3 | NP_003153.2 | 15 | 1863 | ||
XM_005264519.4 | XP_005264576.1 | 14 | 1887 | |||
XM_011533073.2 | XP_011531375.1 | 16 | 2085 | |||
GenBank mRNA | AJ223814.1 | - | 15 | 1863 | ||
AK026934.1 | - | 12 | 1434 | |||
BC106878.2 | - | 16 | 1780 | |||
BC106879.2 | - | 15 | 1715 | 40027865 |