Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000460.3 | NP_000451.1 | 6 | 1026 | ||
NM_001177597.2 | NP_001171068.1 | 6 | 1014 | |||
NM_001177598.2 | NP_001171069.1 | 6 | 1009 | |||
NM_001289997.1 | NP_001276926.1 | 7 | 910 | |||
NM_001289998.1 | NP_001276927.1 | 7 | 1293 | |||
NM_001290003.1 | NP_001276932.1 | 7 | 1293 | |||
NM_001290022.1 | NP_001276951.1 | 7 | 1281 | |||
NM_001290026.1 | NP_001276955.1 | 7 | 1276 | |||
NM_001290027.1 | NP_001276956.1 | 8 | 1177 | |||
NM_001290028.1 | NP_001276957.1 | 6 | 990 | |||
XM_017007107.1 | XP_016862596.1 | 8 | 1165 | |||
GenBank mRNA | AB102890.1 | - | 4 | 730 | ||
AK311002.1 | - | 4 | 420 | |||
BC130322.1 | - | 6 | 946 | 40146353 | ||
BC136338.1 | - | 6 | 958 | 9020325 | ||
BC143982.1 | - | 6 | 946 | 9052500 | ||
D32047.1 | - | 5 | 848 | |||
L33410.1 | - | 6 | 962 | |||
L36052.1 | - | 4 | 747 | |||
U11025.1 | - | 5 | 782 | |||
U59493.1 | - | 4 | 747 | |||
U59494.1 | - | 4 | 747 | |||
U59495.1 | - | 5 | 630 |