Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199427.1 | NP_001186356.1 | 4, 5 | 611 | ||
NM_003300.3 | NP_003291.2 | 4, 5 | 611 | |||
NM_145725.2 | NP_663777.1 | 5, 6 | 750 | |||
NM_145726.2 | NP_663778.1 | 5, 6 | 750 | |||
XM_011537116.2 | XP_011535418.1 | 5, 6 | 726 | |||
XM_011537117.2 | XP_011535419.1 | 5, 6 | 726 | |||
XM_011537118.2 | XP_011535420.1 | 5, 6 | 727 | |||
XM_017021617.1 | XP_016877106.1 | 5, 6 | 1170 | |||
XM_017021618.1 | XP_016877107.1 | 5, 6 | 2666 | |||
XM_017021619.1 | XP_016877108.1 | 5, 6 | 2668 | |||
XM_017021620.1 | XP_016877109.1 | 5, 6 | 728 | |||
GenBank mRNA | AK303172.1 | - | 4, 5 | 488 | ||
BC075086.2 | - | 3, 4 | 422 | 30915387 | ||
BC075087.2 | - | 3, 4 | 422 | 30915599 | ||
BX247977.1 | - | 5, 6 | 596 | |||
L38509.1 | - | 4, 5 | 533 | |||
U19260.1 | - | 4, 5 | 547 | |||
U21092.1 | - | 4, 5 | 614 |