Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP5K1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278253.1 | NP_001265182.1 | 11 | 1594 | ||
NM_003558.3 | NP_003549.1 | 12 | 1751 | |||
XM_005252261.3 | XP_005252318.1 | 11 | 1791 | |||
XM_005252262.4 | XP_005252319.1 | 12 | 1547 | |||
XM_006717300.2 | XP_006717363.1 | 12 | 1657 | |||
XM_006717301.1 | XP_006717364.1 | 12 | 1468 | |||
XM_011519082.2 | XP_011517384.1 | 12 | 4418 | |||
XM_011519083.2 | XP_011517385.1 | 12 | 1572 | |||
XM_011519084.2 | XP_011517386.1 | 13 | 1691 | |||
XM_017015188.1 | XP_016870677.1 | 12 | 1601 | |||
XM_017015189.1 | XP_016870678.1 | 13 | 1716 | |||
XM_017015190.1 | XP_016870679.1 | 11 | 1537 | |||
XM_017015191.1 | XP_016870680.1 | 12 | 1657 | |||
GenBank mRNA | AK292734.1 | - | 12 | 1468 | ||
AK295587.1 | - | 11 | 1349 | |||
BC030587.2 | - | 13 | 1585 | 4823087 | ||
U78579.1 | - | 5 | 542 | |||
U78580.1 | - | 3 | 143 | |||
U78581.1 | - | 6 | 828 |