Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003608.3 | NP_003599.2 | 2 | 1077 | ||
GenBank mRNA | BC035633.1 | - | 2 | 1049 | 5218256 | |
BC071715.1 | - | 2 | 1011 | 5180030 | ||
EU883574.1 | - | 1 | 519 |