Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004701.3 | NP_004692.1 | 3 | 401 | ||
GenBank mRNA | AB020981.1 | - | 3 | 286 | ||
AF002822.1 | - | 3 | 236 | |||
AF087910.1 | - | 3 | 343 | |||
AK001404.1 | - | 3 | 365 | |||
AK222611.1 | - | 3 | 334 | |||
AL080146.1 | - | 3 | 354 | |||
AM851054.1 | - | 3 | 217 | |||
BC105086.1 | - | 3 | 349 | 8144089 | ||
BC105112.1 | - | 3 | 349 | 8144115 | ||
BT019458.1 | - | 3 | 217 | |||
BU934242.1 | - | 3 | 402 | |||
CR533527.1 | - | 3 | 217 |