Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC4A7 SIRNA Assays ›
Gene Symbol:
SLC4A7
Gene Name:
solute carrier family 4 member 7
Gene Aliases:
NBC2, NBC3, NBCN1, SBC2, SLC4A6
Chromosome Location:
Chr.3: 27372721 - 27484420 on Build GRCh38
UniGene ID:
Hs.250072
Species:
Human
siRNA ID
HSS114147
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC4A7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258379.1
NP_001245308.1
20, 21
3098
NM_001258380.1
NP_001245309.1
20, 21
2860
NM_001321103.1
NP_001308032.1
21, 22
3482
NM_001321104.1
NP_001308033.1
21, 22
3443
NM_001321105.1
NP_001308034.1
21, 22
3443
NM_001321106.1
NP_001308035.1
21, 22
3431
NM_001321107.1
NP_001308036.1
20, 21
3098
NM_001321108.1
NP_001308037.1
20, 21
3110
NM_003615.4
NP_003606.3
21, 22
3232
NR_135541.1
20, 21
3256
NR_135542.1
19, 20
2923
NR_135543.1
19, 20
2923
NR_135544.1
20, 21
3256
XM_005265598.4
XP_005265655.1
20, 21
3364
XM_005265600.4
XP_005265657.1
20, 21
3326
XM_006713421.2
XP_006713484.1
21, 22
3470
XM_011534256.2
XP_011532558.1
21, 22
3743
XM_011534258.1
XP_011532560.1
21, 22
3446
XM_011534261.2
XP_011532563.1
21, 22
3708
XM_011534263.2
XP_011532565.1
20, 21
3035
XM_011534265.1
XP_011532567.1
20, 21
3074
XM_017007526.1
XP_016863015.1
22, 23
3367
XM_017007527.1
XP_016863016.1
20, 21
3059
XM_017007528.1
XP_016863017.1
14, 15
1983
XM_017007529.1
XP_016863018.1
14, 15
1965
GenBank mRNA
AF047033.1
-
21, 22
3232
AF053755.1
-
17, 18
2538
AF089726.1
-
20, 21
2860
CR627428.1
-
13, 14
1814
EU499349.1
-
20, 21
3025
EU934246.1
-
21, 22
3358
EU934247.1
-
21, 22
3370
EU934248.1
-
21, 22
3409
EU934249.1
-
20, 21
3025
EU934250.1
-
21, 22
3370
FJ178574.1
-
20, 21
3183
FJ178575.1
-
19, 20
2850
FJ178576.1
-
19, 20
2850
GU354307.1
-
20, 21
3183
GU354308.1
-
20, 21
3037
GU354309.1
-
19, 20
2862
GU354310.1
-
19, 20
2862
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
inorganic anion transport
bicarbonate transport
sodium ion transmembrane transport
regulation of intracellular pH
auditory receptor cell development
anion transmembrane transport
inorganic anion exchanger activity
anion transmembrane transporter activity
sodium:bicarbonate symporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-bqghl:80/10.239.167.51:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0