Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3PXD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014631.2 | NP_055446.2 | 14 | 2738 | ||
XM_005270294.4 | XP_005270351.1 | 15 | 2871 | |||
XM_005270295.4 | XP_005270352.1 | 14 | 2825 | |||
XM_005270297.3 | XP_005270354.1 | 9 | 4102 | |||
XM_005270298.4 | XP_005270355.1 | 9 | 2905 | |||
XM_006718079.3 | XP_006718142.1 | 16 | 2968 | |||
XM_006718080.3 | XP_006718143.1 | 10 | 2999 | |||
XM_011540385.2 | XP_011538687.1 | 15 | 2922 | |||
XM_011540386.2 | XP_011538688.1 | 10 | 4198 | |||
XM_011540387.2 | XP_011538689.1 | 6 | 5844 | |||
XM_017016969.1 | XP_016872458.1 | 15 | 2889 | |||
XM_017016970.1 | XP_016872459.1 | 14 | 2705 | |||
GenBank mRNA | AB007878.1 | - | 12 | 2488 | ||
AK056469.1 | - | 13 | 2461 | |||
AK294454.1 | - | 9 | 2278 | |||
AK294736.1 | - | 9 | 2316 | |||
AK295660.1 | - | 9 | 2382 | |||
AK304828.1 | - | 9 | 2294 | |||
AK315983.1 | - | 9 | 2321 |