Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATG13 SIRNA Assays ›
Gene Symbol:
ATG13
Gene Name:
autophagy related 13
Gene Aliases:
KIAA0652, PARATARG8
Chromosome Location:
Chr.11: 46617276 - 46676019 on Build GRCh38
UniGene ID:
Hs.127403
Species:
Human
siRNA ID
HSS114713
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142673.2
NP_001136145.1
15
1852
NM_001205119.1
NP_001192048.1
15
1895
NM_001205120.1
NP_001192049.1
14
1796
NM_001205121.1
NP_001192050.1
14
1736
NM_001205122.1
NP_001192051.1
12
1603
NM_014741.4
NP_055556.2
14
1741
XM_005253262.2
XP_005253319.1
16
1691
XM_005253263.2
XP_005253320.1
16
1982
XM_005253265.2
XP_005253322.1
17
2188
XM_005253266.2
XP_005253323.1
16
1917
XM_005253268.2
XP_005253325.1
14
1294
XM_006718394.2
XP_006718457.1
17
2038
XM_006718395.2
XP_006718458.1
17
1973
XM_006718396.2
XP_006718459.1
16
2072
XM_011520493.1
XP_011518795.1
18
2175
XM_011520496.2
XP_011518798.1
15
1950
XM_011520499.2
XP_011518801.1
15
1564
XM_011520500.1
XP_011518802.1
17
2011
XM_011520501.1
XP_011518803.1
16
1939
XM_011520502.1
XP_011518804.1
16
1912
XM_011520503.1
XP_011518805.1
15
1828
XM_017018599.1
XP_016874088.1
17
2123
XM_017018600.1
XP_016874089.1
16
1664
XM_017018601.1
XP_016874090.1
15
1839
XM_017018602.1
XP_016874091.1
16
2089
XM_017018603.1
XP_016874092.1
17
2206
XM_017018604.1
XP_016874093.1
15
1883
XM_017018605.1
XP_016874094.1
16
1874
XM_017018606.1
XP_016874095.1
15
1818
XM_017018607.1
XP_016874096.1
13
1195
XM_017018608.1
XP_016874097.1
15
1565
XM_017018609.1
XP_016874098.1
15
1856
XM_017018610.1
XP_016874099.1
14
1740
XM_017018611.1
XP_016874100.1
15
1791
XM_017018612.1
XP_016874101.1
15
1978
XM_017018613.1
XP_016874102.1
14
1772
XM_017018614.1
XP_016874103.1
15
1763
XM_017018615.1
XP_016874104.1
14
1862
XM_017018616.1
XP_016874105.1
13
1656
XM_017018617.1
XP_016874106.1
15
1861
XM_017018618.1
XP_016874107.1
17
2177
XM_017018619.1
XP_016874108.1
15
1962
XM_017018620.1
XP_016874109.1
15
2022
XM_017018621.1
XP_016874110.1
14
1906
XM_017018622.1
XP_016874111.1
14
1863
GenBank mRNA
AB014552.1
-
14
1476
AK023867.1
-
14
1474
AK223603.1
-
15
1571
AK289641.1
-
15
1399
AK293109.1
-
14
1440
AK294110.1
-
12
1316
AK296526.1
-
7
683
AK315925.1
-
7
610
BC001331.1
-
14
1170
3458359
BC002378.2
-
14
1440
2961127
BC006191.2
-
14
1482
3936851
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
macroautophagy
mitophagy in response to mitochondrial depolarization
positive regulation of protein targeting to mitochondrion
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-sjrww:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0