Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SFI1 SIRNA Assays ›
Gene Symbol:
SFI1
Gene Name:
SFI1 centrin binding protein
Gene Aliases:
PISD, PPP1R139, hSfi1p
Chromosome Location:
Chr.22: 31496139 - 31618554 on Build GRCh38
UniGene ID:
Hs.62209
Species:
Human
siRNA ID
HSS114791
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SFI1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007467.2
NP_001007468.1
10
1600
NM_001258325.1
NP_001245254.1
9
1528
NM_001258326.1
NP_001245255.1
8
1350
NM_001258327.1
NP_001245256.1
8
1354
NM_014775.3
NP_055590.2
10
1600
XM_005261868.2
XP_005261925.1
10
1598
XM_006724390.2
XP_006724453.1
8
1352
XM_011530570.2
XP_011528872.1
10
1598
XM_011530571.2
XP_011528873.1
10
1594
XM_011530574.2
XP_011528876.1
10
1275
XM_011530575.2
XP_011528877.1
10
1598
XM_011530576.2
XP_011528878.1
9
1526
XM_011530577.2
XP_011528879.1
9
1522
XM_011530579.2
XP_011528881.1
10
1275
XM_011530580.2
XP_011528882.1
10
1598
XM_011530581.2
XP_011528883.1
10
1275
XM_011530582.2
XP_011528884.1
8
1352
XM_011530583.2
XP_011528885.1
8
1348
XM_011530587.2
XP_011528889.1
2
309
XM_017029139.1
XP_016884628.1
10
1275
XM_017029140.1
XP_016884629.1
9
1526
XM_017029141.1
XP_016884630.1
10
1275
XM_017029142.1
XP_016884631.1
9
1526
XM_017029143.1
XP_016884632.1
10
1594
XM_017029144.1
XP_016884633.1
10
1598
XM_017029145.1
XP_016884634.1
9
1526
XM_017029146.1
XP_016884635.1
10
1275
XM_017029147.1
XP_016884636.1
9
1526
XM_017029148.1
XP_016884637.1
9
1522
XM_017029149.1
XP_016884638.1
9
1526
XM_017029150.1
XP_016884639.1
9
1522
XM_017029151.1
XP_016884640.1
10
1594
XM_017029152.1
XP_016884641.1
8
1352
XM_017029153.1
XP_016884642.1
8
1348
XM_017029154.1
XP_016884643.1
9
1522
XM_017029155.1
XP_016884644.1
8
1352
XM_017029156.1
XP_016884645.1
8
1352
XM_017029157.1
XP_016884646.1
6
761
XM_017029158.1
XP_016884647.1
10
1594
XM_017029159.1
XP_016884648.1
9
1203
XM_017029161.1
XP_016884650.1
8
1029
XM_017029162.1
XP_016884651.1
8
1029
GenBank mRNA
AB011114.2
-
10
1464
AK055147.1
-
2
191
AK292408.1
-
10
1176
AK293987.1
-
9
1141
AK303362.1
-
8
985
AL708501.1
-
1
150
BC110814.1
-
8
951
6500257
BC129926.1
-
6
893
40124134
BC129945.1
-
9
1280
40124130
DA202472.1
-
2
191
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
negative regulation of phosphatase activity
protein binding
phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0