Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPM2AIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014805.3 | NP_055620.1 | 1 | 1363 | ||
GenBank mRNA | AB018309.1 | - | 1 | 1252 | ||
AF059751.1 | - | 1 | 1234 | |||
AK002069.1 | - | 1 | 176 | |||
AY178832.1 | - | 1 | 1363 | |||
BC064696.1 | - | 1 | 1227 | 30345499 |