Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAGI2 SIRNA Assays ›
Gene Symbol:
MAGI2
Gene Name:
membrane associated guanylate kinase, WW and PDZ domain containing 2
Gene Aliases:
ACVRIP1, AIP-1, AIP1, ARIP1, MAGI-2, SSCAM
Chromosome Location:
Chr.7: 78017055 - 79453805 on Build GRCh38
UniGene ID:
Hs.603842
Species:
Human
siRNA ID
HSS114904
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAGI2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301128.1
NP_001288057.1
10
1960
NM_012301.3
NP_036433.2
10
1960
XM_011516718.1
XP_011515020.1
10
2191
XM_011516719.2
XP_011515021.1
10
5608
XM_011516720.2
XP_011515022.1
10
10598
XM_011516726.2
XP_011515028.1
7
1264
XM_011516728.1
XP_011515030.1
6
800
XM_017012840.1
XP_016868329.1
11
2320
XM_017012841.1
XP_016868330.1
11
2317
XM_017012842.1
XP_016868331.1
11
2320
XM_017012843.1
XP_016868332.1
11
2320
XM_017012844.1
XP_016868333.1
11
2320
XM_017012845.1
XP_016868334.1
10
2191
XM_017012846.1
XP_016868335.1
10
2191
XM_017012847.1
XP_016868336.1
11
8557
XM_017012848.1
XP_016868337.1
9
5476
XM_017012849.1
XP_016868338.1
9
5476
XM_017012850.1
XP_016868339.1
11
2320
XM_017012851.1
XP_016868340.1
11
2320
XM_017012852.1
XP_016868341.1
11
2320
GenBank mRNA
AB014605.2
-
10
1941
AF038563.1
-
10
1960
AK297353.1
-
10
1945
AK297580.1
-
9
1519
AK298988.1
-
10
1835
AK308685.1
-
6
781
AK309012.1
-
6
802
BC150277.1
-
10
1941
9007255
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of receptor internalization
planar cell polarity pathway involved in axis elongation
negative regulation of cell proliferation
positive regulation of neuron projection development
negative regulation of cell migration
positive regulation of phosphoprotein phosphatase activity
negative regulation of activin receptor signaling pathway
nerve growth factor signaling pathway
receptor clustering
protein heterooligomerization
negative regulation of protein kinase B signaling
SMAD protein signal transduction
mitotic cell cycle arrest
glomerular visceral epithelial cell development
neuroligin clustering involved in postsynaptic membrane assembly
cellular response to nerve growth factor stimulus
positive regulation of synaptic vesicle clustering
signal transducer activity
protein binding
phosphatase binding
receptor signaling complex scaffold activity
beta-1 adrenergic receptor binding
SMAD binding
type II activin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-bqghl:80/10.239.167.51:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0