Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EXOG SIRNA Assays ›
Gene Symbol:
EXOG
Gene Name:
endo/exonuclease (5'-3'), endonuclease G-like
Gene Aliases:
ENDOGL1, ENDOGL2, ENGL, ENGL-a, ENGL-b, ENGLA, ENGLB
Chromosome Location:
Chr.3: 38496127 - 38526305 on Build GRCh38
UniGene ID:
Hs.517897
Species:
Human
siRNA ID
HSS115059
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EXOG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145464.1
NP_001138936.1
3, 4
481
NM_001318954.1
NP_001305883.1
5, 6
601
NM_001318955.1
NP_001305884.1
6, 7
699
NM_001318956.1
NP_001305885.1
5, 6
698
NM_001318957.1
NP_001305886.1
7, 8
837
NM_001318958.1
NP_001305887.1
7, 8
755
NM_001318959.1
NP_001305888.1
6, 7
793
NM_005107.3
NP_005098.2
4, 5
631
NR_134938.1
4, 5
537
XM_005265633.3
XP_005265690.1
4, 5
477
XM_005265635.3
XP_005265692.1
4, 5
580
XM_006713442.3
XP_006713505.1
5, 6
863
XM_006713444.3
XP_006713507.1
4, 5
474
XM_017007584.1
XP_016863073.1
4, 5
3792
XM_017007585.1
XP_016863074.1
5, 6
2602
XM_017007586.1
XP_016863075.1
4, 5
2527
XM_017007587.1
XP_016863076.1
6, 7
682
XM_017007588.1
XP_016863077.1
6, 7
927
XM_017007589.1
XP_016863078.1
7, 8
994
XM_017007590.1
XP_016863079.1
5, 6
484
XM_017007591.1
XP_016863080.1
5, 6
481
XM_017007592.1
XP_016863081.1
6, 7
593
XM_017007593.1
XP_016863082.1
5, 6
403
GenBank mRNA
AB020523.1
-
4, 5
562
AK023235.1
-
4, 5
538
AK289602.1
-
7, 8
662
AK290107.1
-
5, 6
601
AK301067.1
-
3, 4
410
AK308814.1
-
4, 5
465
AK315814.1
-
6, 7
568
BC104212.1
-
5, 6
627
40031649
BC104213.1
-
6, 7
630
40031650
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
nucleic acid phosphodiester bond hydrolysis
nucleic acid binding
endonuclease activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-86647:80/10.239.162.121:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0