Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306070.1 | NP_001292999.1 | 4 | 1216 | ||
NM_144778.3 | NP_659002.1 | 4 | 1209 | |||
NM_207304.2 | NP_997187.1 | 4 | 1209 | |||
XM_005254018.2 | XP_005254075.1 | 4 | 1494 | |||
XM_005254019.2 | XP_005254076.1 | 4 | 1494 | |||
XM_005254021.2 | XP_005254078.1 | 4 | 1494 | |||
XM_005254022.2 | XP_005254079.1 | 4 | 1494 | |||
XM_006719912.2 | XP_006719975.1 | 4 | 1494 | |||
XM_011521045.2 | XP_011519347.1 | 4 | 1101 | |||
XM_017020308.1 | XP_016875797.1 | 4 | 1101 | |||
XM_017020309.1 | XP_016875798.1 | 4 | 1085 | |||
XM_017020310.1 | XP_016875799.1 | 4 | 1047 | |||
XM_017020311.1 | XP_016875800.1 | 4 | 1085 | |||
GenBank mRNA | AF061261.1 | - | 4 | 1151 | ||
AF491866.1 | - | 4 | 1150 | |||
AK291727.1 | - | 4 | 1156 | |||
AK315844.1 | - | 4 | 1132 | |||
AY101770.1 | - | 3 | 444 | |||
BC020418.1 | - | 4 | 1033 | |||
BC020430.1 | - | 3 | 662 | |||
BC032426.1 | - | 4 | 1140 | |||
BC104038.1 | - | 4 | 1137 | 40026585 | ||
BC104039.1 | - | 4 | 1137 | 40026586 | ||
BC104040.1 | - | 4 | 1137 | 40026588 | ||
CR749802.1 | - | 5 | 1168 |