Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006369.4 | NP_006360.3 | 5 | 1556 | ||
XM_017000074.1 | XP_016855563.1 | 5 | 1582 | |||
GenBank mRNA | AF370430.1 | - | 5 | 1456 | ||
AK125827.1 | - | 4 | 2833 | |||
AK223420.1 | - | 2 | 918 | |||
AK291283.1 | - | 5 | 1539 | |||
AK296436.1 | - | 5 | 1375 | |||
AK297596.1 | - | 5 | 1570 | |||
BC004953.1 | - | 2 | 792 | |||
BC092479.1 | - | 5 | 1366 | |||
BC101522.1 | - | 5 | 1546 | 8069028 | ||
BC101524.1 | - | 5 | 1546 | 8069030 | ||
BX537366.1 | - | 5 | 1466 | |||
X86018.1 | - | 2 | 768 |