Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261412.1 | NP_001248341.1 | 7 | 759 | ||
NM_001261413.1 | NP_001248342.1 | 6 | 753 | |||
NM_006400.4 | NP_006391.1 | 6 | 768 | |||
XM_011537761.2 | XP_011536063.1 | 7 | 720 | |||
GenBank mRNA | AK055682.1 | - | 6 | 631 | ||
AK096242.1 | - | 8 | 627 | |||
AK098794.1 | - | 7 | 613 | |||
AK222693.1 | - | 6 | 631 | |||
AK292364.1 | - | 8 | 646 | |||
AK314705.1 | - | 6 | 610 | |||
AY189155.1 | - | 7 | 491 | |||
BC000718.2 | - | 6 | 617 | 3504109 | ||
BC009468.1 | - | 6 | 543 | 3613836 | ||
BC014083.2 | - | 6 | 591 | 4303142 | ||
FJ224315.1 | - | 6 | 578 | |||
U50733.1 | - | 6 | 578 |