Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLU7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006425.4 | NP_006416.3 | 8 | 953 | ||
XM_005265793.3 | XP_005265850.1 | 8 | 1300 | |||
XM_011534418.2 | XP_011532720.1 | 9 | 1003 | |||
GenBank mRNA | AF101074.1 | - | 7 | 798 | ||
AK024875.1 | - | 8 | 873 | |||
AK314564.1 | - | 8 | 873 | |||
BC010634.1 | - | 8 | 879 | 3871882 | ||
CA976106.1 | - | 6 | 544 |