Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
siRNA ID
HSS116292
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
9, 10
1030
NM_001034955.1
NP_001030127.1
9, 10
1030
NM_001034956.1
NP_001030128.1
9, 10
823
NM_001034957.1
NP_001030129.1
6, 7
538
NM_001290294.1
NP_001277223.1
11, 12
1217
NM_001290295.1
NP_001277224.1
9, 10
752
NM_001290296.1
NP_001277225.1
8, 9
1003
NM_001290297.1
NP_001277226.1
7, 8
565
NM_001290298.1
NP_001277227.1
7, 8
634
NM_006434.2
NP_006425.2
6, 7
538
NM_015385.3
NP_056200.1
10, 11
1121
NM_024991.2
NP_079267.1
10, 11
848
XM_006717589.1
XP_006717652.1
10, 11
1143
XM_006717593.1
XP_006717656.1
8, 9
703
XM_011539140.1
XP_011537442.1
12, 13
1887
XM_011539150.1
XP_011537452.1
12, 13
1887
XM_011539155.1
XP_011537457.1
10, 11
1789
XM_011539167.1
XP_011537469.1
9, 10
1072
XM_017015500.1
XP_016870989.1
12, 13
1887
XM_017015501.1
XP_016870990.1
12, 13
1887
XM_017015502.1
XP_016870991.1
12, 13
1887
XM_017015503.1
XP_016870992.1
12, 13
1887
XM_017015504.1
XP_016870993.1
10, 11
801
XM_017015505.1
XP_016870994.1
12, 13
1887
XM_017015506.1
XP_016870995.1
10, 11
801
XM_017015507.1
XP_016870996.1
12, 13
1887
XM_017015508.1
XP_016870997.1
10, 11
801
XM_017015509.1
XP_016870998.1
9, 10
730
XM_017015510.1
XP_016870999.1
12, 13
1887
XM_017015511.1
XP_016871000.1
10, 11
801
XM_017015512.1
XP_016871001.1
8, 9
703
XM_017015513.1
XP_016871002.1
8, 9
703
XM_017015514.1
XP_016871003.1
8, 9
703
XM_017015515.1
XP_016871004.1
11, 12
1170
XM_017015516.1
XP_016871005.1
10, 11
1143
XM_017015517.1
XP_016871006.1
11, 12
1170
XM_017015518.1
XP_016871007.1
10, 11
1099
XM_017015519.1
XP_016871008.1
9, 10
1072
XM_017015520.1
XP_016871009.1
11, 12
1170
XM_017015521.1
XP_016871010.1
10, 11
1099
XM_017015522.1
XP_016871011.1
10, 11
1143
XM_017015523.1
XP_016871012.1
10, 11
1099
XM_017015524.1
XP_016871013.1
9, 10
1072
XM_017015525.1
XP_016871014.1
9, 10
730
XM_017015526.1
XP_016871015.1
10, 11
801
XM_017015527.1
XP_016871016.1
9, 10
730
XM_017015528.1
XP_016871017.1
9, 10
774
XM_017015529.1
XP_016871018.1
10, 11
801
XM_017015530.1
XP_016871019.1
10, 11
801
XM_017015531.1
XP_016871020.1
9, 10
730
XM_017015532.1
XP_016871021.1
9, 10
774
XM_017015533.1
XP_016871022.1
8, 9
703
XM_017015534.1
XP_016871023.1
10, 11
801
XM_017015535.1
XP_016871024.1
9, 10
730
XM_017015536.1
XP_016871025.1
9, 10
730
XM_017015537.1
XP_016871026.1
9, 10
774
XM_017015538.1
XP_016871027.1
9, 10
774
XM_017015539.1
XP_016871028.1
8, 9
703
XM_017015540.1
XP_016871029.1
8, 9
703
GenBank mRNA
AB037717.1
-
10, 11
780
AF136380.1
-
5, 6
493
AF136381.1
-
7, 8
958
AF330623.1
-
6, 7
520
AF356525.1
-
9, 10
1041
AF356526.1
-
9, 10
1041
AF356527.1
-
9, 10
834
AJ489942.1
-
6, 7
589
AK300407.1
-
9, 10
705
AK304753.1
-
10, 11
1764
AL117472.1
-
10, 11
1079
AM260536.1
-
11, 12
1175
BC042612.1
-
8, 9
1145
BC152463.1
-
10, 11
780
8862599
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kt7sl:80/10.239.164.98:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0