Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SDCCAG8 SIRNA Assays ›
Gene Symbol:
SDCCAG8
Gene Name:
serologically defined colon cancer antigen 8
Gene Aliases:
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
Chromosome Location:
Chr.1: 243255328 - 243500092 on Build GRCh38
UniGene ID:
Hs.591530
Species:
Human
siRNA ID
HSS116682
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SDCCAG8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_006642.3
NP_006633.1
9
1115
XM_005273013.4
XP_005273070.1
8
947
XM_005273018.1
XP_005273075.1
8
954
XM_005273021.4
XP_005273078.1
9
887
XM_005273022.3
XP_005273079.1
6
736
XM_005273023.4
XP_005273080.1
9
1792
XM_006711727.3
XP_006711790.1
9
1001
XM_006711728.3
XP_006711791.1
8
872
XM_011544021.2
XP_011542323.1
10
1097
XM_011544022.2
XP_011542324.1
10
1172
XM_011544023.2
XP_011542325.1
10
1097
XM_011544024.2
XP_011542326.1
10
1097
XM_011544026.2
XP_011542328.1
10
1097
XM_011544028.2
XP_011542330.1
8
872
XM_011544029.2
XP_011542331.1
10
1097
XM_011544030.2
XP_011542332.1
2
1648
XM_017000103.1
XP_016855592.1
10
1097
XM_017000104.1
XP_016855593.1
8
947
XM_017000105.1
XP_016855594.1
9
1102
XM_017000106.1
XP_016855595.1
9
1083
XM_017000107.1
XP_016855596.1
10
999
XM_017000108.1
XP_016855597.1
12
1546
XM_017000109.1
XP_016855598.1
9
1001
GenBank mRNA
AF039690.1
-
3
286
AF161348.1
-
9
1092
AF250731.1
-
9
946
AK297105.1
-
5
534
AK299212.1
-
9
1064
AK299249.1
-
6
707
BC032454.1
-
9
1058
5174799
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
neuron migration
establishment of cell polarity
tube formation
centrosome organization
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0