Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007027.3 | NP_008958.2 | 21 | 3703 | ||
XM_005247076.3 | XP_005247133.1 | 21 | 4175 | |||
XM_017005636.1 | XP_016861125.1 | 21 | 3773 | |||
XM_017005637.1 | XP_016861126.1 | 18 | 3227 | |||
GenBank mRNA | AB019397.1 | - | 21 | 3650 | ||
AK302584.1 | - | 21 | 3669 | |||
BC126209.1 | - | 20 | 3593 | 8991925 | ||
BC151237.1 | - | 21 | 3656 | 8860504 | ||
D87448.1 | - | 21 | 3656 | |||
Y18557.1 | - | 6 | 796 |