Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031693.2 | NP_001026863.1 | 1 | 240 | ||
NM_001036645.1 | NP_001031722.1 | 1 | 240 | |||
NM_001036646.1 | NP_001031723.1 | 1 | 240 | |||
NR_027404.1 | 1 | 240 | ||||
XM_011540547.2 | XP_011538849.1 | 1 | 1210 | |||
XM_011540548.2 | XP_011538850.1 | 1 | 135 | |||
GenBank mRNA | AF126163.1 | - | 1 | 62 | ||
AF126164.1 | - | 1 | 221 | |||
BC010922.1 | - | 1 | 218 | 4064190 | ||
BG773961.1 | - | 1 | 240 | |||
BI868263.1 | - | 1 | 240 | |||
BQ009559.1 | - | 4 | 626 |