Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRAK1 SIRNA Assays ›
Gene Symbol:
TRAK1
Gene Name:
trafficking kinesin protein 1
Gene Aliases:
MILT1, OIP106
Chromosome Location:
Chr.3: 42013802 - 42225890 on Build GRCh38
UniGene ID:
Hs.535711
Species:
Human
siRNA ID
HSS117874
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRAK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042646.2
NP_001036111.1
12, 13
1650
NM_001265608.1
NP_001252537.1
12, 13
1650
NM_001265609.1
NP_001252538.1
11, 12
1513
NM_001265610.1
NP_001252539.1
11, 12
1513
NM_014965.4
NP_055780.2
11, 12
1453
XM_005264956.2
XP_005265013.1
12, 13
1458
XM_005264958.2
XP_005265015.1
11, 12
1513
XM_005264960.3
XP_005265017.1
12, 13
1458
XM_005264962.3
XP_005265019.1
12, 13
1458
XM_005264963.3
XP_005265020.1
12, 13
1458
XM_006713029.2
XP_006713092.1
12, 13
1458
XM_006713030.2
XP_006713093.1
12, 13
1458
XM_006713031.3
XP_006713094.1
12, 13
1458
XM_011533489.2
XP_011531791.1
12, 13
1458
XM_017005906.1
XP_016861395.1
14, 15
2071
XM_017005907.1
XP_016861396.1
11, 12
1435
XM_017005908.1
XP_016861397.1
11, 12
1513
XM_017005909.1
XP_016861398.1
13, 14
1956
XM_017005910.1
XP_016861399.1
11, 12
1513
XM_017005911.1
XP_016861400.1
13, 14
1956
XM_017005912.1
XP_016861401.1
12, 13
1458
XM_017005913.1
XP_016861402.1
12, 13
1458
GenBank mRNA
AB028965.1
-
12, 13
1650
AK000754.1
-
11, 12
1429
AK294221.1
-
11, 12
1479
AK295480.1
-
12, 13
1688
AK295848.1
-
12, 13
1557
AK316260.1
-
12, 13
1547
AL713787.1
-
11, 12
1417
BC015922.1
-
11, 12
1440
4562916
BF527591.1
-
3, 4
419
BX647199.1
-
11, 12
1453
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
protein O-linked glycosylation
protein targeting
endosome to lysosome transport
protein binding
GABA receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-lhtlm:80/10.239.167.245:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0