Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DOPEY1 SIRNA Assays ›
Gene Symbol:
DOPEY1
Gene Name:
dopey family member 1
Gene Aliases:
DOP1, KIAA1117, dJ202D23.2
Chromosome Location:
Chr.6: 83067650 - 83171351 on Build GRCh38
UniGene ID:
Hs.520246
Species:
Human
siRNA ID
HSS118065
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DOPEY1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199942.1
NP_001186871.1
31, 32
6468
NM_015018.3
NP_055833.2
31, 32
6495
XM_011535619.2
XP_011533921.1
30, 31
6460
XM_017010559.1
XP_016866048.1
31, 32
6490
XM_017010560.1
XP_016866049.1
30, 31
6418
XM_017010561.1
XP_016866050.1
31, 32
6442
XM_017010562.1
XP_016866051.1
30, 31
6391
XM_017010563.1
XP_016866052.1
31, 32
6469
XM_017010564.1
XP_016866053.1
31, 32
6469
XM_017010565.1
XP_016866054.1
31, 32
6463
XM_017010566.1
XP_016866055.1
31, 32
6463
XM_017010567.1
XP_016866056.1
30, 31
6391
XM_017010568.1
XP_016866057.1
30, 31
6460
XM_017010569.1
XP_016866058.1
29, 30
6377
XM_017010570.1
XP_016866059.1
30, 31
6391
XM_017010571.1
XP_016866060.1
30, 31
6385
XM_017010573.1
XP_016866062.1
31, 32
6490
XM_017010574.1
XP_016866063.1
31, 32
6490
GenBank mRNA
AB029040.2
-
14, 15
3516
AK027030.1
-
11, 12
2262
AL050080.1
-
3, 4
224
BC048342.1
-
5, 6
508
BC144544.1
-
31, 32
6366
9053071
BC150619.1
-
31, 32
6366
9056989
BX648667.1
-
31, 32
6493
Back To Top
More Information
Gene Ontology Categories:
Function(s)
protein transport
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-7dvbj:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0