Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAST2 SIRNA Assays ›
Gene Symbol:
MAST2
Gene Name:
microtubule associated serine/threonine kinase 2
Gene Aliases:
MAST205, MTSSK
Chromosome Location:
Chr.1: 45786992 - 46036124 on Build GRCh38
UniGene ID:
Hs.319481
Species:
Human
siRNA ID
HSS118242
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAST2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001319245.1
NP_001306174.1
17, 18
2325
NM_001324320.1
NP_001311249.1
18, 19
2352
NM_001324321.1
NP_001311250.1
17, 18
2438
NM_015112.2
NP_055927.2
17, 18
2325
XM_005270655.3
XP_005270712.1
16, 17
2045
XM_005270656.4
XP_005270713.1
16, 17
1914
XM_006710477.3
XP_006710540.1
15, 16
1722
XM_011541059.2
XP_011539361.1
19, 20
2559
XM_011541061.2
XP_011539363.1
19, 20
2559
XM_011541062.2
XP_011539364.1
18, 19
2538
XM_011541063.1
XP_011539365.1
19, 20
2123
XM_011541064.2
XP_011539366.1
19, 20
2120
XM_011541065.1
XP_011539367.1
19, 20
2222
XM_011541067.2
XP_011539369.1
15, 16
2036
XM_011541068.1
XP_011539370.1
14, 15
1653
XM_011541069.2
XP_011539371.1
19, 20
2559
XM_017000752.1
XP_016856241.1
17, 18
2552
XM_017000753.1
XP_016856242.1
16, 17
1886
XM_017000754.1
XP_016856243.1
15, 16
1865
XM_017000755.1
XP_016856244.1
16, 17
2357
GenBank mRNA
AB018350.2
-
6, 7
637
AB047005.1
-
17, 18
2325
AK027123.1
-
15, 16
1864
AK096519.1
-
16, 17
1649
BC006166.1
-
9, 10
993
BC015816.2
-
9, 10
993
BC065499.1
-
17, 18
2299
5499830
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
cytoskeleton organization
peptidyl-serine phosphorylation
intracellular signal transduction
regulation of interleukin-12 biosynthetic process
spermatid differentiation
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
microtubule binding
phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-jmz56:80/10.239.163.191:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0