Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PHLDB1 SIRNA Assays ›
Gene Symbol:
PHLDB1
Gene Name:
pleckstrin homology like domain family B member 1
Gene Aliases:
LL5A
Chromosome Location:
Chr.11: 118606296 - 118658038 on Build GRCh38
UniGene ID:
Hs.504062
Species:
Human
siRNA ID
HSS118328
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PHLDB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144758.2
NP_001138230.1
11
2755
NM_001144759.2
NP_001138231.1
11
2755
NM_015157.3
NP_055972.1
12
2978
XM_005271468.3
XP_005271525.1
11
3524
XM_005271469.3
XP_005271526.1
11
3524
XM_005271470.3
XP_005271527.1
11
3524
XM_005271471.3
XP_005271528.1
11
3524
XM_005271472.3
XP_005271529.1
11
3524
XM_005271473.3
XP_005271530.1
11
3524
XM_005271474.4
XP_005271531.1
11
3524
XM_005277680.3
XP_005277737.1
12
3650
XM_006718796.3
XP_006718859.1
12
2839
XM_006718797.3
XP_006718860.1
12
3653
XM_006718798.3
XP_006718861.1
12
3653
XM_006718799.3
XP_006718862.1
12
3653
XM_006718800.3
XP_006718863.1
12
3653
XM_006718801.3
XP_006718864.1
12
3653
XM_006718802.3
XP_006718865.1
12
3653
XM_006718803.3
XP_006718866.1
12
3653
XM_011542703.2
XP_011541005.1
12
3653
XM_011542704.2
XP_011541006.1
12
3653
XM_011542705.2
XP_011541007.1
12
3653
XM_011542706.2
XP_011541008.1
12
3653
XM_011542707.2
XP_011541009.1
11
3524
XM_011542708.2
XP_011541010.1
12
2842
XM_011542709.2
XP_011541011.1
12
3152
XM_011542710.2
XP_011541012.1
12
3653
XM_011542711.2
XP_011541013.1
9
2559
XM_011542713.2
XP_011541015.1
9
2553
XM_011542715.2
XP_011541017.1
6
2031
XM_017017407.1
XP_016872896.1
11
3521
XM_017017408.1
XP_016872897.1
12
3653
XM_017017409.1
XP_016872898.1
12
3653
XM_017017410.1
XP_016872899.1
11
3524
GenBank mRNA
AB014538.2
-
11
2732
AB094090.1
-
12
2978
AB177859.1
-
6
1859
AK027510.1
-
3
384
AK074070.2
-
11
2704
AK074918.1
-
5
591
AK125783.1
-
13
1982
AL832373.1
-
11
2712
BC013031.2
-
5
741
BC098586.1
-
11
2732
30342198
BX648176.1
-
6
907
CR749783.1
-
12
2891
Back To Top
More Information
Gene Ontology Categories:
Function(s)
regulation of gastrulation
regulation of epithelial to mesenchymal transition
regulation of microtubule cytoskeleton organization
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f2ldd:80/10.239.165.14:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0