Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADGRL2 SIRNA Assays ›
Gene Symbol:
ADGRL2
Gene Name:
adhesion G protein-coupled receptor L2
Gene Aliases:
CIRL2, CL2, LEC1, LPHH1, LPHN2
Chromosome Location:
Chr.1: 81306155 - 81992738 on Build GRCh38
UniGene ID:
Hs.24212
Species:
Human
siRNA ID
HSS118486
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADGRL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001297704.1
NP_001284633.1
13
2567
NM_001297705.1
NP_001284634.1
12
2602
NM_001297706.1
NP_001284635.1
12
2602
NM_012302.3
NP_036434.1
12
2602
XM_005270666.4
XP_005270723.1
13
3362
XM_005270668.2
XP_005270725.1
12
2619
XM_006710485.3
XP_006710548.1
14
3378
XM_006710488.2
XP_006710551.1
14
2672
XM_006710489.3
XP_006710552.1
14
2563
XM_017000782.1
XP_016856271.1
18
3246
XM_017000783.1
XP_016856272.1
15
2657
XM_017000784.1
XP_016856273.1
14
2562
XM_017000785.1
XP_016856274.1
14
2669
XM_017000786.1
XP_016856275.1
14
2673
XM_017000787.1
XP_016856276.1
14
2584
XM_017000788.1
XP_016856277.1
14
2560
XM_017000789.1
XP_016856278.1
13
2544
XM_017000790.1
XP_016856279.1
15
2598
XM_017000791.1
XP_016856280.1
13
2662
XM_017000792.1
XP_016856281.1
13
2558
XM_017000793.1
XP_016856282.1
15
2600
XM_017000794.1
XP_016856283.1
15
2599
XM_017000795.1
XP_016856284.1
15
2598
XM_017000796.1
XP_016856285.1
13
2622
XM_017000797.1
XP_016856286.1
12
2528
XM_017000798.1
XP_016856287.1
12
2590
GenBank mRNA
AB018329.1
-
8
1113
AF104266.1
-
12
2602
AF104938.1
-
12
2602
AF104939.1
-
12
2602
AJ131581.1
-
13
2566
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
latrotoxin receptor activity
carbohydrate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0