Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323614.1 | NP_001310543.1 | 15 | 2347 | ||
NM_001323615.1 | NP_001310544.1 | 14 | 2209 | |||
NM_001323616.1 | NP_001310545.1 | 16 | 2479 | |||
NM_001323617.1 | NP_001310546.1 | 16 | 2314 | |||
NM_015327.2 | NP_056142.2 | 15 | 2286 | |||
XM_017000842.1 | XP_016856331.1 | 11 | 2232 | |||
XM_017000843.1 | XP_016856332.1 | 14 | 2214 | |||
GenBank mRNA | AB029012.2 | - | 15 | 2288 | ||
AB085691.1 | - | 15 | 2214 | |||
AK027486.1 | - | 2 | 74 | |||
AL137738.1 | - | 4 | 388 | |||
AY168922.1 | - | 15 | 2140 | |||
AY336728.1 | - | 15 | 2137 | |||
BC007453.2 | - | 4 | 733 | |||
BC038296.1 | - | 15 | 2287 | 4824487 |