Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301167.1 | NP_001288096.1 | 6 | 630 | ||
NM_013252.2 | NP_037384.1 | 7 | 675 | |||
XM_011515995.2 | XP_011514297.1 | 5 | 562 | |||
XM_017011915.1 | XP_016867404.1 | 6 | 633 | |||
XM_017011916.1 | XP_016867405.1 | 7 | 697 | |||
XM_017011917.1 | XP_016867406.1 | 6 | 628 | |||
GenBank mRNA | AF139768.1 | - | 7 | 626 | ||
AJ271684.1 | - | 7 | 675 | |||
AK292766.1 | - | 7 | 664 | |||
AK295200.1 | - | 6 | 567 | |||
AK307095.1 | - | 5 | 493 | |||
AK313703.1 | - | 7 | 664 | |||
BC093714.1 | - | 6 | 499 | 7939559 | ||
BC112099.1 | - | 6 | 499 | 8327567 | ||
BC113099.1 | - | 6 | 548 | 40076725 | ||
BC143392.1 | - | 5 | 426 |